missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine, TRC

Product Code. 30135787
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135787 50 g 50g
30134152 25 g 25g
17928089 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135787 Supplier Toronto Research Chemicals Supplier No. TRCD41624850G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-95-2
Molecular Formula C12 H14 N4
Molecular Weight (g/mol) 214.27
Synonym 3,3',4,4'-Biphenyltetramine (6CI,8CI), 3,3',4,4'-Tetraaminobiphenyl, 3,3',4,4'-Tetraaminodiphenyl, 3,3'-Diaminobenzidine, 3,4,3',4'-Tetraaminobiphenyl, Betazoid DAB, Biphenyl-3,3',4,4'-tetraamine, DAB, NSC 76152
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine
SMILES Nc1ccc(cc1N)c2ccc(N)c(N)c2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2
Formula Weight 214.1218
Chemical Name or Material 3,3'-Diaminobenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.