accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 913835-43-5

BOHNNNCH3HOOHHCl

CAS RN 913835-43-5

IUPAC Name: {4-[(4-methylpiperazin-1-yl)carbamoyl]phenyl}boronic acid hydrochloride
Synonyms: 4-[(4-methylpiperazin-1-yl)carbamoyl]phenylboronic acid hydrochloride
Molecular Weight (g/mol): 299.56
Molecular Formula: C12H19BClN3O3
InChi Key: DPTCRDJSYQTDGH-UHFFFAOYSA-N
SMILES: Cl.CN1CCN(CC1)NC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results