accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 914225-70-0

H3COFI

CAS RN 914225-70-0

IUPAC Name: 1-(5-fluoro-2-iodophenyl)ethan-1-one
Synonyms: 1-(5-fluoro-2-iodophenyl)ethanone
Molecular Weight (g/mol): 264.04
Molecular Formula: C8H6FIO
InChi Key: NRLSRIONJVBZDT-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(I)C=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results