accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 917022-26-5

ONHOO

CAS RN 917022-26-5

IUPAC Name: benzyl N-(4-formylcyclohexyl)carbamate
Synonyms: benzyl N-(4-formylcyclohexyl)carbamate
Molecular Weight (g/mol): 261.32
Molecular Formula: C15H19NO3
InChi Key: SOIRQISQWQDGSN-UHFFFAOYSA-N
SMILES: O=CC1CCC(CC1)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)