accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 918643-56-8

BOOH3CCH3H3CH3CNONCH3CH3

CAS RN 918643-56-8

IUPAC Name: dimethyl(3-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxy}propyl)amine
Synonyms: dimethyl(3-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxy}propyl)amine
Molecular Weight (g/mol): 306.21
Molecular Formula: C16H27BN2O3
InChi Key: QWJUJXPYTLOKRL-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results