accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 923977-15-5

IUPAC Name: 3-(3,5-dibromophenyl)propanoic acid
Synonyms: 3-(3,5-dibromophenyl)propanoic acid
Molecular Weight (g/mol): 307.97
Molecular Formula: C9H8Br2O2
InChi Key: AMKWPMUHANVQSZ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(Br)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results