missing translation for 'onlineSavingsMsg'
Learn More

3-(3,5-Dibromophenyl)propionic acid, 97%

Product Code. 15460458
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460458 5 g 5g
15450458 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460458 Supplier Thermo Scientific Alfa Aesar Supplier No. H54012.06
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 923977-15-5 | C9H8Br2O2 | 307.969 g/mol

3-(3,5-Dibromophenyl)propionic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3,5-Dibromophenyl)propionic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923977-15-5
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.969
MDL Number MFCD11973898
InChI Key AMKWPMUHANVQSZ-UHFFFAOYSA-N
Synonym 3-3,5-dibromophenyl propanoic acid, 3-3,5-dibromophenyl propionic acid, 3,5-dibromobenzenepropanoic acid, 3-3,5-dibromo-phenyl-propionic acid
PubChem CID 16038256
IUPAC Name 3-(3,5-dibromophenyl)propanoic acid
SMILES C1=C(C=C(C=C1Br)Br)CCC(=O)O

Specifications

Melting Point 62°C
Quantity 5 g
Beilstein 10537616
Formula Weight 307.97
Percent Purity 97%
Chemical Name or Material 3-(3,5-Dibromophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.