accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OBrBr

CAS RN 923977-18-8

IUPAC Name: 5,7-dibromo-2,3-dihydro-1H-inden-1-one
Synonyms: 5,7-dibromo-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 289.95
Molecular Formula: C9H6Br2O
InChi Key: WLRPGDOWFWBUAE-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C2C(=O)CCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results