CAS RN 926921-67-7
IUPAC Name:
1-{4-[(6-methylpyrazin-2-yl)oxy]phenyl}methanamine
Synonyms:
1-{4-[(6-methylpyrazin-2-yl)oxy]phenyl}methanamine
Molecular Weight (g/mol):
215.26
Molecular Formula:
C12H13N3O
InChi Key:
JNWHMYUBZLJKJT-UHFFFAOYSA-N
SMILES:
CC1=CN=CC(OC2=CC=C(CN)C=C2)=N1
1
–
1
of
1
results
1
4-[(6-Methylpyrazin-2-yl)oxy]benzylamine, 97%, Thermo Scientific™
CAS: 926921-67-7 Molecular Formula: C12H13N3O Molecular Weight (g/mol): 215.256 MDL Number: MFCD09879937 InChI Key: JNWHMYUBZLJKJT-UHFFFAOYSA-N Synonym: 4-6-methylpyrazin-2-yl oxy benzylamine,4-6-methylpyrazin-2-yl oxy phenyl methanamine,1-4-6-methylpyrazin-2-yl oxy phenyl methanamine,4-6-methylpyrazin-2-yloxy phenyl methylamine PubChem CID: 24229693 IUPAC Name: [4-(6-methylpyrazin-2-yl)oxyphenyl]methanamine SMILES: CC1=CN=CC(=N1)OC2=CC=C(C=C2)CN