missing translation for 'onlineSavingsMsg'
Learn More

4-[(6-Methylpyrazin-2-yl)oxy]benzylamine, 97%, Thermo Scientific™

Product Code. 10708845
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10708845 1 g Amber Glass Bottle 1g
10618624 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10708845 Supplier Thermo Scientific Maybridge Supplier No. CC62713DA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-67-7
Molecular Formula C12H13N3O
Molecular Weight (g/mol) 215.256
MDL Number MFCD09879937
InChI Key JNWHMYUBZLJKJT-UHFFFAOYSA-N
Synonym 4-6-methylpyrazin-2-yl oxy benzylamine, 4-6-methylpyrazin-2-yl oxy phenyl methanamine, 1-4-6-methylpyrazin-2-yl oxy phenyl methanamine, 4-6-methylpyrazin-2-yloxy phenyl methylamine
PubChem CID 24229693
IUPAC Name [4-(6-methylpyrazin-2-yl)oxyphenyl]methanamine
SMILES CC1=CN=CC(=N1)OC2=CC=C(C=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.25
Percent Purity 95%
Chemical Name or Material 4-[(6-Methylpyrazin-2-yl)oxy]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.