accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CO(E)

CAS RN 927-63-9

IUPAC Name: (2E)-3-(dimethylamino)prop-2-enal
Synonyms: (2E)-3-(dimethylamino)prop-2-enal
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: RRLMPLDPCKRASL-ONEGZZNKSA-N
SMILES: CN(C)\C=C\C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results