accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSOO

CAS RN 93-11-8

IUPAC Name: naphthalene-2-sulfonyl chloride
Synonyms: 2-naphthalenesulfonyl chloride
Molecular Weight (g/mol): 226.67
Molecular Formula: C10H7ClO2S
InChi Key: OPECTNGATDYLSS-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (8)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (5)
  • (5)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (3)
  • (5)
  • (5)
  • (8)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (5)
  • (3)
  • (2)