missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene-2-sulfonyl Chloride, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Naphthalene-2-sulfonyl Chloride |
| CAS | 93-11-8 |
| Molecular Formula | C10H7ClO2S |
| Quantity | 100 mg |
| Synonym | 2-Naphthalenylsulfonyl Chloride, 2-Naphthylsulfonyl Chloride, NSC 133893, Naphthalene-2-sulfonic Acid Chloridebeta-Naphthalenesulfochloride |
| InChI Formula | InChI=1S/C10H7ClO2S/c11-14(12,13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H |
| SMILES | ClS(=O)(=O)c1ccc2ccccc2c1 |
| IUPAC Name | naphthalene-2-sulfonyl chloride |
| Molecular Weight (g/mol) | 226.68 |
| Formula Weight | 225.9855 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction