accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 930-28-9

IUPAC Name: chlorocyclopentane
Synonyms: cyclopentane, chloro-
Molecular Weight (g/mol): 104.58
Molecular Formula: C5H9Cl
InChi Key: NDTCXABJQNJPCF-UHFFFAOYSA-N
SMILES: ClC1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)