missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclopentane, 99%

Product Code. 11415987
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415987 500 g 500g
11405987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415987 Supplier Thermo Scientific Alfa Aesar Supplier No. B25023.36
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 930-28-9 | C5H9Cl | 104.577 g/mol

Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. 760 mg/L.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-28-9
Molecular Formula C5H9Cl
Molecular Weight (g/mol) 104.577
MDL Number MFCD00001360
InChI Key NDTCXABJQNJPCF-UHFFFAOYSA-N
Synonym cyclopentane, chloro, cyclopentyl chloride, chloroclopentane, cyclopentylchloride, unii-to3f41cl9a, 1-chlorocyclopentane, to3f41cl9a, cyclopentylchlorid, chloro-cyclopentan, chlorocyclopenetane
PubChem CID 70252
IUPAC Name chlorocyclopentane
SMILES C1CCC(C1)Cl

Specifications

Melting Point -94°C
Density 1.003
Boiling Point 113°C to 114°C
Flash Point 15°C (59°F)
Refractive Index 1.4512
Quantity 500 g
UN Number UN1993
Beilstein 1900557
Solubility Information Soluble in water. 760mg/L.
Formula Weight 104.58
Percent Purity 99%
Chemical Name or Material Chlorocyclopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.