CAS RN 93118-03-7
IUPAC Name:
2-chloro-3-(trifluoromethyl)benzaldehyde
Synonyms:
2-chloro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
208.56
Molecular Formula:
C8H4ClF3O
InChi Key:
KUNCMOAFNYLOSC-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC(C=O)=C1Cl
1
–
2
of
2
results
1
2-Chloro-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
CAS: 93118-03-7 Molecular Formula: C8H4ClF3O Molecular Weight (g/mol): 208.564 MDL Number: MFCD00792414 InChI Key: KUNCMOAFNYLOSC-UHFFFAOYSA-N Synonym: 2-chloro-3-trifluoromethyl benzaldehyde,2-chloro-3-trifluoromethyl-benzaldehyde,benzaldehyde, 2-chloro-3-trifluoromethyl,pubchem2880,acmc-20aib9,ksc486s7h,timtec-bb sbb003469,attercop-chm at119733,2-chloro-3 trifluoromethyl-benzaldehyde PubChem CID: 2736672 IUPAC Name: 2-chloro-3-(trifluoromethyl)benzaldehyde SMILES: C1=CC(=C(C(=C1)C(F)(F)F)Cl)C=O