missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

Product Code. 10234954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10234954 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10234954 Supplier Thermo Scientific Acros Supplier No. 398430010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 93118-03-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 43°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H4ClF3O
Linear Formula ClC6H3(CF3)CHO
MDL Number MFCD00792414
Quantity 1 g
Synonym 2-chloro-3-trifluoromethyl benzaldehyde, 2-chloro-3-trifluoromethyl-benzaldehyde, benzaldehyde, 2-chloro-3-trifluoromethyl, pubchem2880, acmc-20aib9, ksc486s7h, timtec-bb sbb003469, attercop-chm at119733, 2-chloro-3 trifluoromethyl-benzaldehyde
InChI Key KUNCMOAFNYLOSC-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)C(F)(F)F)Cl)C=O
IUPAC Name 2-chloro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol) 208.57
PubChem CID 2736672
Formula Weight 208.57
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2-chloro-3-(trifluoromethyl)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3-(trifluoromethyl)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.