accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 932-16-1

CH3NH3CO

CAS RN 932-16-1

IUPAC Name: 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one
Synonyms: 2-acetyl-1-methylpyrrole
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: NZFLWVDXYUGFAV-UHFFFAOYSA-N
SMILES: CN1C=CC=C1C(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results