CAS RN 933-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-4,6-dichloro-s-triazine, TRC
CAS: 933-20-0 Molecular Formula: C3 H2 Cl2 N4 Molecular Weight (g/mol): 164.98 IUPAC Name: 4,6-dichloro-1,3,5-triazin-2-amine SMILES: Nc1nc(Cl)nc(Cl)n1