missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloro-s-triazine, TRC

Product Code. 30135274
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135274 5 g 5g
17990825 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135274 Supplier Toronto Research Chemicals Supplier No. TRCA6047505G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933-20-0
Molecular Formula C3 H2 Cl2 N4
Molecular Weight (g/mol) 164.98
IUPAC Name 4,6-dichloro-1,3,5-triazin-2-amine
SMILES Nc1nc(Cl)nc(Cl)n1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H2Cl2N4/c4-1-7-2(5)9-3(6)8-1/h(H2,6,7,8,9)
Formula Weight 163.9657
Chemical Name or Material 2-Amino-4,6-dichloro-s-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.