CAS RN 933702-55-7
CAS RN 933702-55-7
IUPAC Name:
2-chloropyrimidine-5-carbaldehyde
Synonyms:
2-chloropyrimidine-5-carbaldehyde
Molecular Weight (g/mol):
142.54
Molecular Formula:
C5H3ClN2O
InChi Key:
LJYQVOPFBNMTKJ-UHFFFAOYSA-N
SMILES:
ClC1=NC=C(C=O)C=N1
1
–
1
of
1
results
1
2-chloropyrimidine-5-carboxaldehyde, 97%
CAS: 933702-55-7 Molecular Formula: C5H3ClN2O Molecular Weight (g/mol): 142.54 MDL Number: MFCD10696891 InChI Key: LJYQVOPFBNMTKJ-UHFFFAOYSA-N Synonym: 2-chloropyrimidine-5-carboxyaldehyde,2-chloropyrimidine-5-carboxaldehyde,2-chloro-5-pyrimidinecarboxaldehyde,pubchem23896,2-chloro-5-formyl pyrimidine,chloropyrimidine-5-carbaldehyde,2-chloro-pyrimidine-5-carbaldehyde,2-chloranylpyrimidine-5-carbaldehyde,5-pyrimidinecarboxaldehyde, 2-chloro PubChem CID: 21698350 IUPAC Name: 2-chloropyrimidine-5-carbaldehyde SMILES: ClC1=NC=C(C=O)C=N1