accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 938-73-8

CH3ONH2O

CAS RN 938-73-8

IUPAC Name: 2-ethoxybenzamide
Synonyms: 2-ethoxybenzamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: SBNKFTQSBPKMBZ-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C(N)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)