missing translation for 'onlineSavingsMsg'
Learn More

Ethenzamide, LoGiCal

Product Code. 30459984
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459984 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459984 Supplier LoGiCal Supplier No. LGCFOR3063.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Ethenzamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethenzamide
CAS 938-73-8
Analyte or Component Names Ethenzamide
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O2
Certifications/Compliance ISO 9001
Synonym Benzamide, 2-ethoxy-, Benzamide, o-ethoxy- (6 CI,7 CI,8 CI), 2-Ethoxybenzamide, Etamide, Ethbenzamide, Ethenzamid, Ethenzamide, Ethosalicyl, Etocil, Etosalicil, Etosalicyl, Eusal, Katagrippe, Lucamide, NSC 28787, Pirosolvina, Protopyrin, Trancalgyl, o-Ethoxybenzamide
InChI Formula InChI=1 S/C9H11NO2/c1-2-12-8-6-4-3-5-7(8)9(10)11/h3-6 H,2H2,1H3,(H2,10,11)
SMILES CCOc1ccccc1C(=O)N
IUPAC Name 2-ethoxybenzamide
Industry Type Forensics
Molecular Weight (g/mol) 165.19
Quantity 10 mg
Formula Weight 165.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.