accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHN

CAS RN 942-24-5

IUPAC Name: methyl 1H-indole-3-carboxylate
Synonyms: methyl 1H-indole-3-carboxylate
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: QXAUTQFAWKKNLM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)