accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 944251-24-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
Synonyms: 2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
Molecular Weight (g/mol): 388.38
Molecular Formula: C15H24N4O8
InChi Key: OIGKWPIMJCPGGD-UHFFFAOYSA-N
SMILES: [N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results