missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%

Product Code. 15440397
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440397 100 mg 100mg
15430397 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440397 Supplier Thermo Scientific Alfa Aesar Supplier No. J64834.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 944251-24-5 | C15H24N4O8 | 388.38 g/mol

N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.

Solubility
Soluble in dimethyl sulfoxide and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944251-24-5
Molecular Formula C15H24N4O8
Molecular Weight (g/mol) 388.38
MDL Number MFCD13184948
InChI Key OIGKWPIMJCPGGD-UHFFFAOYSA-N
Synonym Azido-PEG4-NHS ester
IUPAC Name 2,5-dioxopyrrolidin-1-yl 1-azido-3,6,9,12-tetraoxapentadecan-15-oate
SMILES [N-]=[N+]=NCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O

Specifications

Color Colorless to Yellow
Quantity 100 mg
Solubility Information Soluble in dimethyl sulfoxide and acetonitrile.
Formula Weight 388.37
Physical Form Liquid
Chemical Name or Material N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.