accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

S

CAS RN 95-15-8

IUPAC Name: 1-benzothiophene
Synonyms: benzothiophene 2,3-benzothiophene benzothiofuran thianaphtene benzo(b)thiophene benzo(B)thiophene
Molecular Weight (g/mol): 134.2
Molecular Formula: C8H6S
InChi Key: FCEHBMOGCRZNNI-UHFFFAOYSA-N
SMILES: S1C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results