missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Standard for ASTM D5623: Multi-component standard containing 14 sulfur species @ 100 μg/g (as Sulfur) in Base Fuel

Product Code. 30324121
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324121 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324121 Supplier VHG Labs Supplier No. VHGMSX142
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Benzothiophene∣1-Butanethiol∣1-Propanethiol∣2-Propanethiol∣Bromothiophene∣Diethyl disulfide∣Diethylsulfide∣Dimethyl sulfide∣Diphenyl sulfide∣Ethanethiol∣Methanethiol∣tert.-Butanethiol∣Thiophene∣Thiophenol
CAS 95-15-8, 109-79-5, 107-03-9, 75-33-2, 110-81-6, 352-93-2
Analyte or Component Names 1-Benzothiophene∣1-Butanethiol∣1-Propanethiol∣2-Propanethiol∣Bromothiophene∣Diethyl disulfide∣Diethylsulfide∣Dimethyl sulfide∣Diphenyl sulfide∣Ethanethiol∣Methanethiol∣tert.-Butanethiol∣Thiophene∣Thiophenol
Shipping Condition Room Temperature
Solution Type Single Solution
Solvent or Matrix Base Fuel
Industry Type Petrochemical Testing
Quantity 2 mL
Concentration or Composition (by Analyte or Components) Methanethiol 98.6 μg/g∣Ethanethiol 99.6 μg/g∣Dimethyl sulfide 99.9 μg/g∣1-Benzothiophene 100 μg/g∣1-Butanethiol 100 μg/g∣1-Propanethiol 100 μg/g∣tert.-Butanethiol 100 μg/g∣Thiophene 100 μg/g∣Thiophenol 100 μg/g∣2-Propanethiol 101 μg/g∣Bromothiophene 101 μg/g∣Diethyl disulfide 101 μg/g∣Diethylsulfide 101 μg/g∣Diphenyl sulfide 101 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.