accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3

CAS RN 95-92-1

IUPAC Name: diethyl oxalate
Synonyms: ethyl oxalate
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: WYACBZDAHNBPPB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)OCC
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (5)