CAS RN 950912-80-8
CAS RN 950912-80-8
IUPAC Name:
4-[1-cyclohexyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]pyrimidin-2-amine dihydrochloride
Synonyms:
4-[3-cyclohexyl-5-(4-fluorophenyl)imidazol-4-yl]pyrimidin-2-amine dihydrochloride
Molecular Weight (g/mol):
410.32
Molecular Formula:
C19H22Cl2FN5
InChi Key:
PSNKGVAXBSAHCH-UHFFFAOYSA-N
SMILES:
Cl.Cl.NC1=NC=CC(=N1)C1=C(N=CN1C1CCCCC1)C1=CC=C(F)C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ PF 670462
CAS: 950912-80-8 Molecular Formula: C19H22Cl2FN5
2
PF 670462, TRC
CAS: 950912-80-8 Molecular Formula: C19H22Cl2FN5 Molecular Weight (g/mol): 410.32 Synonym: PF 670462,PF 670462 IUPAC Name: 4-[3-cyclohexyl-5-(4-fluorophenyl)imidazol-4-yl]pyrimidin-2-amine;dihydrochloride SMILES: Cl.Cl.Nc1nccc(n1)c2c(ncn2C3CCCCC3)c4ccc(F)cc4