missing translation for 'onlineSavingsMsg'
Learn More

PF 670462, TRC

Product Code. 30414035
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414035 10 mg
30412477 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414035 Supplier Toronto Research Chemicals Supplier No. TRCP29396010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 950912-80-8
Molecular Formula C19H22Cl2FN5
Molecular Weight (g/mol) 410.32
Synonym PF 670462
IUPAC Name 4-[3-cyclohexyl-5-(4-fluorophenyl)imidazol-4-yl]pyrimidin-2-amine;dihydrochloride
SMILES Cl.Cl.Nc1nccc(n1)c2c(ncn2C3CCCCC3)c4ccc(F)cc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H20FN5.2ClH/c20-14-8-6-13(7-9-14)17-18(16-10-11-22-19(21)24-16)25(12-23-17)15-4-2-1-3-5-15;;/h6-12,15H,1-5H2,(H2,21,22,24);2*1H
Formula Weight 409.1236
Chemical Name or Material 4-[1-Cyclohexyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]-2-pyrimidinamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.