accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95333-14-5

IUPAC Name: 1-benzofuran-7-carbaldehyde
Synonyms: 1-benzofuran-7-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: RGPUSZZTRKTMNA-UHFFFAOYSA-N
SMILES: O=CC1=C2OC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results