CAS RN 95596-35-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,5-Pentane-1,1,5,5-d4-diamine, TRC
CAS: 95596-35-3 Molecular Formula: C5 D4 H10 N2 Molecular Weight (g/mol): 106.2 Synonym: 1,1,5,5-tetradeuteriopentane-1,5-diamine IUPAC Name: 1,5-Pentane-1,1,5,5-d4-diamine (9CI) SMILES: [2H]C([2H])(N)CCCC([2H])([2H])N
2
1,5-Pentane-1,1,5,5-d4-diamine, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.