accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 957060-88-7

IUPAC Name: (3-chloro-2-methoxypyridin-4-yl)boronic acid
Synonyms: 3-chloro-2-methoxypyridin-4-ylboronic acid
Molecular Weight (g/mol): 187.39
Molecular Formula: C6H7BClNO3
InChi Key: YQNAXOGPSUVHNU-UHFFFAOYSA-N
SMILES: COC1=NC=CC(B(O)O)=C1Cl
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (1)