accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 95973-51-6

H3COOOCH3

CAS RN 95973-51-6

IUPAC Name: 4-ethoxyphenyl 4-(4-pentylcyclohexyl)benzoate
Synonyms: 4-ethoxyphenyl 4-(4-pentylcyclohexyl)benzoate
Molecular Weight (g/mol): 394.56
Molecular Formula: C26H34O3
InChi Key: BOJBRLBJBXWHNZ-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)C(=O)OC1=CC=C(OCC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results