accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3

CAS RN 96-37-7

IUPAC Name: methylcyclopentane
Synonyms: methylcyclopentane
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: GDOPTJXRTPNYNR-UHFFFAOYSA-N
SMILES: CC1CCCC1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)