missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentane, TRC

Product Code. 17970875
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970875 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970875 Supplier Toronto Research Chemicals Supplier No. TRCM3383035ML
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 96-37-7
Molecular Formula C6 H12
Quantity 5 mL
Synonym Methylcyclopentane, 1-Methyl-cyclopentanecarbaldehyde, Methycyclopentane, NSC 24836
InChI Formula InChI=1S/C6H12/c1-6-4-2-3-5-6/h6H,2-5H2,1H3
SMILES CC1CCCC1
IUPAC Name methylcyclopentane
Molecular Weight (g/mol) 84.16
Formula Weight 84.0939
Chemical Name or Material Methylcyclopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.