accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-00-0

IUPAC Name: (furan-2-yl)methanol
Synonyms: furfuryl alcohol 2-furane-methanol furfuryl alkohol furylcarbinol furfural alcohol 2-furfuryl alcohol furfuranol
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: XPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILES: OCC1=CC=CO1
Molecular Weight (g/mol) 
  • (8)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (8)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (7)
  • (1)