Learn More
Furfuryl Alcohol, TRC
Product Code. 30133782
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133782 250 g 250g
30133537 50 g 50g
18053240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133782 Supplier Toronto Research Chemicals Supplier No. TRCF864220250G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-00-0
Molecular Formula C5 H6 O2
Molecular Weight (g/mol) 98.1
Synonym 2-Hydroxymethylfuran, 2-Furylmethanol, 2-Furfurylalcohol, 2-Furanmethanol, Furfuryl alcohol (8CI), 2-(Hydroxymethyl)furan, 2-Furancarbinol, 2-Furanylmethanol, 2-Furfuryl alcohol, 2-Furylcarbinol, 2-Furylmethyl alcohol, 5-Hydroxymethylfuran, Chem-Rez 200, Chem-Rez Fury 454, Furfuralcohol, Furfuranol, Furylcarbinol, NSC 8843, α-Furfuryl alcohol, α-Furylcarbinol
IUPAC Name furan-2-ylmethanol
SMILES OCc1occc1
Specifications
Quantity 250 g
InChI Formula InChI=1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2
Formula Weight 98.0368
Chemical Name or Material Furfuryl Alcohol