accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-59-9

IUPAC Name: 4-methylbenzene-1-sulfonyl chloride
Synonyms: P-toluenesulfonyl chloride
Molecular Weight (g/mol): 190.64
Molecular Formula: C7H7ClO2S
InChi Key: YYROPELSRYBVMQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (3)
Percent Purity 
  • (3)
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (5)
  • (4)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (3)
  • (3)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (4)
  • (5)