missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl Chloride, TRC

Product Code. 30449028
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449028 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449028 Supplier Toronto Research Chemicals Supplier No. TRCT535905500G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Toluenesulfonyl Chloride
CAS 98-59-9
Purity Grade Notes HPLC
Molecular Formula C7H7ClO2S
Quantity 500 g
Synonym p-Toluenesulfonyl Chloride, 4-Methylbenzene-1-sulfonyl Chloride, 4-Methylbenzenesulfonyl Chloride, 4-Methylphenylsulfonyl Chloride, 4-Toluenesulfonyl Chloride, 4-Toluolsulfonyl Chloride, 4-Tosyl Chloride, NSC 175822, Toluenesulfonyl Chloride, Tosyl Chloride, p-Methylbenzenesulfonyl Chloride, p-Methylphenylsulfonyl Chloride, p-Toluenesulfochloride, p-Toluenesulfonic Acid Chloride, p-Toluenesulfonic Chloride, p-Toluenesulphonyl Chloride, p-Tosyl Chloride, TsCl, pTsCl, p-TsCl','Ts-Cl
InChI Formula InChI=1S/C7H7ClO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3
SMILES Cc1ccc(cc1)S(=O)(=O)Cl
IUPAC Name 4-methylbenzenesulfonyl chloride
Molecular Weight (g/mol) 190.65
Formula Weight 189.9855
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.