accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 98-86-2

IUPAC Name: 1-phenylethan-1-one
Synonyms: acetophenone hypnone fema number 2009 usaf EK-496 rcra waste number U004 acetofenon
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: KWOLFJPFCHCOCG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (5)
  • (3)
Boiling Point 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (7)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (3)
  • (5)
  • (6)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)