CAS RN 98619-07-9
IUPAC Name:
1-(2-fluoro-4-hydroxyphenyl)ethan-1-one
Synonyms:
1-(2-fluoro-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
154.14
Molecular Formula:
C8H7FO2
InChi Key:
ZCZIRBNZMFUCOH-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(O)C=C1F
1
–
1
of
1
results
1
2'-Fluoro-4'-hydroxyacetophenone, 97%
CAS: 98619-07-9 Molecular Formula: C8H7FO2 Molecular Weight (g/mol): 154.14 MDL Number: MFCD00142713 InChI Key: ZCZIRBNZMFUCOH-UHFFFAOYSA-N Synonym: 2'-fluoro-4'-hydroxyacetophenone,1-2-fluoro-4-hydroxyphenyl ethanone,4-hydroxy-2-fluoroacetophenone,2-fluoro-4-hydroxyacetophenone,1-2-fluoro-4-hydroxyphenyl-1-ethanone,1-2-fluoro-4-hydroxyphenyl ethan-1-one,1-2-fluoro-4-hydroxy-phenyl ethanone,ethanone, 1-2-fluoro-4-hydroxyphenyl,4'-hydroxy-2'-fluoroacetophenone PubChem CID: 2724912 IUPAC Name: 1-(2-fluoro-4-hydroxyphenyl)ethanone SMILES: CC(=O)C1=CC=C(O)C=C1F