CAS RN 99-27-4
IUPAC Name:
1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate
Synonyms:
1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate
Molecular Weight (g/mol):
209.2
Molecular Formula:
C10H11NO4
InChi Key:
DEKPYXUDJRABNK-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(=CC(N)=C1)C(=O)OC
1
–
1
of
1
results
1
Dimethyl 5-aminoisophthalate, 98%
CAS: 99-27-4 Molecular Formula: C10H11NO4 Molecular Weight (g/mol): 209.201 MDL Number: MFCD00008435 InChI Key: DEKPYXUDJRABNK-UHFFFAOYSA-N PubChem CID: 66831 IUPAC Name: dimethyl 5-aminobenzene-1,3-dicarboxylate SMILES: COC(=O)C1=CC(=CC(=C1)N)C(=O)OC