accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-66-1

H3CCH3OHO

CAS RN 99-66-1

IUPAC Name: 2-propylpentanoic acid
Synonyms: valproic acid n-dpa phl-valproic acid pms-valproic acid E.C. deproic ergenyl delepsine valproate product acido valproico [inn-spanish] sprinkle
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: NIJJYAXOARWZEE-UHFFFAOYSA-N
SMILES: CCCC(CCC)C(O)=O
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)