missing translation for 'onlineSavingsMsg'
Learn More

Valproic acid, Lipomed

Product Code. 30458956
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30458956

Brand: Lipomed LPMVAL169450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Valproic acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valproic acid
CAS 99-66-1
Analyte or Component Names Valproic acid
Shipping Condition Room Temperature
Molecular Formula C8 H16 O2
Synonym Valproic Acid, 2-Propylpentanoic acid, 2-Propylvaleric acid, 4-Heptanecarboxylic acid, 44089, Acetic acid, dipropyl-, DPA, Depakine, Dipropylacetic acid, Ergenyl, Mylproin, NSC 93819, Valproic acid, n-Dipropylacetic acid
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CCCC(CCC)C(=O)O
IUPAC Name 2-propylpentanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 144.21
Quantity 50 mg
Formula Weight 144.115 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.