accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONH2

CAS RN 99-92-3

IUPAC Name: 1-(4-aminophenyl)ethan-1-one
Synonyms: 4-aminoacetophenone
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: GPRYKVSEZCQIHD-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (3)
  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)