accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONH(S)O

CAS RN 99395-88-7

IUPAC Name: (4S)-4-phenyl-1,3-oxazolidin-2-one
Synonyms: (4S)-4-phenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: QDMNNMIOWVJVLY-MRVPVSSYSA-N
SMILES: O=C1N[C@H](CO1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results