missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Phenyl-2-oxazolidinone, TRC

Product Code. 30426748
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426748 25 g -
30410099 5 g 5g
30426526 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426748 Supplier Toronto Research Chemicals Supplier No. TRCP33582025G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99395-88-7
Molecular Formula C9 H9 N O2
Molecular Weight (g/mol) 163.1733
Synonym 2-Oxazolidinone, 4-phenyl-, (4S)-, 2-Oxazolidinone, 4-phenyl-, (S)-, (+)-4-Phenyl-2-oxazolidinone, (4S)-4-Phenyl-1,3-oxazolidin-2-one, (4S)-4-Phenyl-2-oxazolidinone, (4S)-4-Phenyloxazolidin-2-one, (S)-(+)-4-Phenyl-2-oxazolidinone, (S)-4-Phenyl-1,3-oxazolidin-2-one, (S)-4-Phenyl-2-oxazolidinone, (S)-POZ, (S)-POZ
IUPAC Name (4S)-4-phenyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@H](CO1)c2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H9NO2/c11-9-10-8(6-12-9)7-4-2-1-3-5-7/h1-5,8H,6H2,(H,10,11)/t8-/m1/s1
Formula Weight 163.0633
Chemical Name or Material (S)-(+)-4-Phenyloxazolidin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.