CAS RN 994-31-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
243.36
Molecular Formula:
C6H17ClSn
InChi Key:
PFMCXCNQGDRWRV-UHFFFAOYSA-N
SMILES:
Cl.CC[SnH](CC)CC
1
–
1
of
1
results
1
Triethyltin chloride, Thermo Scientific Chemicals
CAS: 994-31-0 Molecular Formula: C6H17ClSn Molecular Weight (g/mol): 243.36 MDL Number: MFCD00040414 InChI Key: PFMCXCNQGDRWRV-UHFFFAOYSA-N Synonym: triethyltin chloride,stannane, chlorotriethyl,chlorotriethyltin,triethylchlorotin,triethylchlorostannane,triethylstannyl chloride,tin, triethyl-, chloride,triethyltin chloride c2h5 3sncl,triethylstannium chloride,triaethylzinnchlorid PubChem CID: 13820 IUPAC Name: chloro(triethyl)stannane SMILES: Cl.CC[SnH](CC)CC